ContactUsLinkComponent
Label Printers, M710
Label Printers, M710
Supplier:  Brady
New Product
Certificates
undefined
Label Printers, M710
Supplier:  Brady
New Product
Specifications
  • Catalog No:
  • BRDY322918
  • BRDY322913
  • BRDY322912
  • BRDY322916
  • BRDY322917
  • BRDY322914
  • BRDY322911
  • BRDY322915
  • Connector type:
  • Bluetooth, Wi-Fi, USB 2.0, USB-C, standalone use
  • Standalone use, USB 2.0, USB-C
  • Standalone use, USB 2.0, USB-C
  • Bluetooth, Wi-Fi, USB 2.0, USB-C, standalone use
  • Bluetooth, Wi-Fi, USB 2.0, USB-C, standalone use
  • Bluetooth, Wi-Fi, USB 2.0, USB-C, standalone use
  • Standalone use, USB 2.0, USB-C
  • Bluetooth, Wi-Fi, USB 2.0, USB-C, standalone use
  • Display:
  • Colour LCD touchscreen
  • Colour LCD touchscreen
  • Colour LCD touchscreen
  • Colour LCD touchscreen
  • Colour LCD touchscreen
  • Colour LCD touchscreen
  • Colour LCD touchscreen
  • Colour LCD touchscreen
  • Electrical:
  • 110 - 240 V
  • 110 - 240 V
  • 110 - 240 V
  • 110 - 240 V
  • 110 - 240 V
  • 110 - 240 V
  • 110 - 240 V
  • 110 - 240 V
  • Memory:
  • 200 MB
  • 200 MB
  • 200 MB
  • 200 MB
  • 200 MB
  • 200 MB
  • 200 MB
  • 200 MB
  • Model:
  • M710-CYRILLIC
  • M710-QWERTY-UK
  • M710-AZERTY
  • M710-QWERTY
  • M710-QWERTY-UK
  • M710-QWERTY-EU
  • M710-QWERTY-EU
  • M710-AZERTY-EU
  • Power supply:
  • AC adaptor, rechargeable battery
  • AC adaptor, rechargeable battery
  • AC adaptor, rechargeable battery
  • AC adaptor, rechargeable battery
  • AC adaptor, rechargeable battery
  • AC adaptor, rechargeable battery
  • AC adaptor, rechargeable battery
  • AC adaptor, rechargeable battery
  • Printer:
  • Monocolour
  • Monocolour
  • Monocolour
  • Monocolour
  • Monocolour
  • Monocolour
  • Monocolour
  • Monocolour
  • Print method:
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Thermal transfer
  • Print resolution:
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • 300 dpi
  • Print speed:
  • 76,20 mm/s
  • 76,20 mm/s
  • 76,20 mm/s
  • 76,20 mm/s
  • 76,20 mm/s
  • 76,20 mm/s
  • 76,20 mm/s
  • 76,20 mm/s
  • L×W×H:
  • 116,75×240,03×345,20 mm
  • 116,75×240,03×345,20 mm
  • 116,75×240,03×345,20 mm
  • 116,75×240,03×345,20 mm
  • 116,75×240,03×345,20 mm
  • 116,75×240,03×345,20 mm
  • 116,75×240,03×345,20 mm
  • 116,75×240,03×345,20 mm