ContactUsLinkComponent
LabFlame+ Bunsen Burner, with Integrated NIMI (AA)
LabFlame+ Bunsen Burner, with Integrated NIMI (AA)
Catalog # BTOLLF8.204.000
Supplier:  BioTool
New Product
LabFlame+ Bunsen Burner, with Integrated NIMI (AA)
Catalog # BTOLLF8.204.000
Supplier:  BioTool
New Product
Specifications
  • Description:
    LabFlame+ bunsen burner, with integrated NIMI (AA)
Specifications
About this item
The LabFlame+  is optimised for use in microbiology laboratories. Its stainless steel housing is made of one piece, free of any gaps or rims and therefore easy to clean and fully UV resistant. Its optimised air-flow system allows a uniform and constant flame. Connectable to any kind of gas sources. Ideal for use in cleanroom workbenches and laboratories.
  • The animated and high-contrast graphic display can be easily read under all lighting conditions due to the blue background illumination
    The self-explanatory, language-less symbol menu facilitates a rapid selection of all functions
    Acoustic signals additionally aid the user
    The precisely adjustable, exact to the second, cooling time reminder aids you in exactly complying with the required cooling period of inoculation loops
    Excellent energy efficiency in accordance with the most recent standards reduces gas and power consumption, increases the burning time
    Can be operated with stationary natural gas and propane / butane gas supplies, cartridge gas or gas cylinders
The exclusive LabFlame+ provides maximum flexibility. Wireless operation using state-of-the-art rechargeable battery technology guarantees independent operation. Two standard rechargeable batteries allow up to 9 hours of continuous operation. This corresponds to approximately 2000 inoculation loop flaming operations. The LabFlame+ has an integrated fast charging function and can be recharged in just 3 hours.
The LabFlame+ can be operated with the touch-free IR-Sensor, button function or foot pedal. For all options, different programs exist that have been developed on the basis of practical experience: flexible start-stop functions, continuous application for up to 2 hours or programs for short flame sterilization - exact to the second. When the burner is activated the display shows the remaining burning time.
The zero-pressure shut-down provides additional safety at the end of work. With it the residual pressure is released from the connection hose and the gas hose's service life is increase.
Fabricated entirely of stainless steel, with fireproof controls and a display, protected by heat resistant glass, the LabFlame+ can withstand extreme lab conditions. 
The unique shape is eye-catching. The low casing makes convenient working possible; the streamlined design reduces the disturbance of the air current above a clean room workbench to a minimum. DualKnob for adjusting gas and air flow, the flame can be precisely regulated. The removable burner head can simply be dismantled into its individual components for in-depth cleaning. Clogging is no longer a problem. Spilled liquids flow off through an integrated drain, and solid substances can be effortlessly removed by opening the burner shaft.