ContactUsLinkComponent
Productainer™ Aegis™ 3D BioProcess Container (BPC), 5 to 14 Film
Productainer™ Aegis™ 3D BioProcess Container (BPC), 5 to 14 Film
Catalog # HYCPSH30966.03
New Product
Productainer™ Aegis™ 3D BioProcess Container (BPC), 5 to 14 Film
Catalog # HYCPSH30966.03
New Product
Specifications
  • Capacity:
    50 L
  • Description:
    Top dispense, C-Flex tubing style
  • Environmentally Preferable:
  • Port:
    4 ports
Specifications
About this item
Productainer 3D BioProcess Containers (BPCs) are used to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products.
  • Film optimized for flexibility and durability - superior stress resistance, puncture resistance, and physical barrier properties to ensure stability of contents
    Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components
Productainer BPCs are constructed with Aegis 5 to 14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form, and function, Productainer BPCs can be customized using the industry's largest component library.
Recommended for: Harvest - collection and storage of bulk product from bioreactors. Storage - sterile storage of media, buffers, and process intermediates. Separation - feeding and receiving of sterile liquids from separation systems. Purification - buffer feeding to purification system, fraction collection and storage. Filling - sterile reservoir for filling systems. Transport and shipping - bulk sterile liquids, buffers, media and process intermediates within and between facilities.