Orders LinkContactUsLinkComponent
Flow Through Cell
Flow Through Cell
Flow through cell, glass body
Catalog # SENK255-75
New Product
Supplier:   SENTEK
Flow Through Cell
Catalog # SENK255-75
Supplier:  SENTEK
New Product
Flow Through Cell
Flow through cell, glass body
Price
108,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Glass flow through cell for 12 mm pH and conductivity electrodes.
Designed for general purpose applications, particularly good for measuring conductivity in pure water.
Caution: Not for pressure applications - only use to drain.
Specifications
  • Description:
    Flow through cell, glass body