Gonyautoxin-1 and Gonyautoxin-4 Solution
Catalog # 55758-0.5ML
Supplier: Merck
New Product
CAS Number:
Specifications
- Concentration:20 μg/g in acetic acid
- Description:Gonyautoxin-1 and Gonyautoxin-4 solution, 0,5 ml
Specifications
About this item
Gonyautoxin-1 and Gonyautoxin-4 solution 20 μg/g in acetic acid (nominal concentration), analytical standard.
Our high-purity standards help to keep the food and cosmetic products safe and allow the manufacturers and laboratories to meet all legal regulations for label claims.