- Capacity:2×2,5 L
- Description:Cleaner and disinfectant for non-invasive medical devices and nonporous surfaces
- Environmentally Preferable:
- Pack type:Box (Traded unit), Bottles (Consumer unit)
Diversey Oxivir Excel combined cleaner and disinfectant, powered by AHP Technology, provides you with the best alternative to your traditional disinfectants by delivering a full virucidal efficacy in just 30 seconds, while being very gentle to surfaces and with the lowest risk to your staff and customer. The active ingredient, hydrogen peroxide, breaks down into environmentally-friendly water and oxygen just minutes after use leaving no residues that might be harmful or change the appearance of the surface.
Full virucidal claim in just 30 sec
Fast kill claims for a broad spectrum of viruses, bacteria and yeasticidal
Superior cleaning with high-performance surfactant system
Fragrance free
No rinsing required
Safer for the environment, as well for the users, since the actives breaks down into water and oxygen as APEs, NPEs and VOCs
Benefits:
- Broad efficacy provides the highest levels of disinfection. Extensively tested against European norms and effective against multiple pathogens of concern, carries a full virucidal claim including Polio, Adeno, and Murine norovirus, considered effective against all enveloped viruses such as: HIV, HBV, HCV, Influenza, Vaccinia and Coronaviruses.
- Safe for users no need PPE. No hazard classification
- Superior cleaning performance and 10x faster than your normal disinfectant
- AHP formulation is gentle on surfaces extending equipment life. No residues.
- Suitable for use in high risk areas in retail, education, healthcare and other sectors
- Improve the disinfection compliance at 35%
Dimension (W×L×H): 138×249×240 mm
Weight: 5,32 kg (Net weight), 5,83 kg (Gross weight)