Mechanical Gloves for Work in Dry Environments
Lieferant: DELTA PLUS
New Product
Zertifikate
Über diesen Artikel
Mechanical gloves ideal for work in dry environments. The anti-slip properties of the polyurethane coating provide good grip.
Knitted coated gloves
Support: Tricot - polyester
Benefits element: Reach
Applications: Public Works, finishing works/craftsmanship, maintenance, automotive, temporary work, water and waste treatment, power energy, storage.
Certifications:
EN 388:2016+A1:2018.
Spezifikationen
- Catalog No:
- DELPVE702P/8
- DELPVE702P/10
- Material:
- Polyester
- Polyester
- Finish:
- Smooth PU palm coating (Single coated)
- Smooth PU palm coating (Single coated)
- Gauge:
- 13
- 13
Spezifikationen
Frequently Bought Together