Tryptic Soy Broth, Dehydrated Culture Media
Artikelnr. 1.04326.5000
Lieferant: Merck
New Product
CAS-Nummer:
Spezifikationen
- Description:Tryptic soy broth - dehydrated culture Media
- Format:Powder
- Storage temperature:15...25 °C
- Application:Pharmaceutical
- pH:7.3 ± 0.2 (25.0 °C)
Spezifikationen
Über diesen Artikel
NutriSelect® prime tryptic soy broth halal is designed as a combination of two peptones, enzymatic digest of casein and of soy-bean, which provides a high nutrition by supplying organic nitrogen, amino acids, and longer-chained peptides. The osmotic balance is maintained by sodium chloride and dipotassium phosphate acts as a buffer. NutriSelect® prime tryptic soy broth halal is used for the isolation and cultivation of a wide range of microorganisms from test samples.
Certifications:
NutriSelect® Prime, EP, USP, JP, ISO, FDA BAM, powder, suitable for use in halal and kosher certified processes.