ContactUsLinkComponent
Tork S4 Antimicrobial Hand Washing Liquid Soap
Tork S4 Antimicrobial Hand Washing Liquid Soap
Catalog # SCAA424709
Supplier:  Essity
New Product
undefined
Tork S4 Antimicrobial Hand Washing Liquid Soap
Catalog # SCAA424709
Supplier:  Essity
New Product
Specifications
  • Packed:
    6× 1000 ml
  • Description:
    Premium liquid soap, refill
Specifications
About this item
Tork antimicrobial hand washing liquid soap S4, dermatologically tested.
  • Help to protect guests - this liquid soap conforms with EN 1499 and EN 14476 and is effective against all enveloped viruses
  • Prove your environmental commitment - this biodegradable liquid soap contains a 100% plant-based active ingredient
  • Be kind to skin - It is dermatologically tested and suitable for frequent use
  • Meet safety standards - this non-flammable soap has no CLP symbols, warning phrases or transport restrictions
  • No scent fragrance, premium quality
    Colour: Transparent
    Compatible with S4 dispensers
Swiftly tackle problem microorganisms with this liquid soap. It’s lactic acid-based formula kills all enveloped viruses, making it ideal for food-handling environments. The hand wash is also made for frequent use, thanks to its dermatologically tested gentle feel.
Its 100% plant-based active ingredient and biodegradable formula means it's a responsible choice. For easy dispensing, use with any Tork skincare dispenser (S4).
Caution:
Use biocides safely. Always read the label and product information before use.