ContactUsLinkComponent
Millipore® Immobilon®-P NOW Bundle Hydrophobic PVDF Transfer Membrane
Millipore® Immobilon®-P NOW Bundle Hydrophobic PVDF Transfer Membrane
Catalog # MILFNWKITIP
Supplier:  Merck
New Product
undefined
Millipore® Immobilon®-P NOW Bundle Hydrophobic PVDF Transfer Membrane
Catalog # MILFNWKITIP
Supplier:  Merck
New Product
Specifications
  • Description:
    NOW bundle, Immobilon®-P hydrophobic PVDF transfer membrane
  • Dimensions:
    85×10000 mm
Specifications
About this item
Hydrophobic immobilon-P PVDF transfer membrane for western blotting using chemiluminescence, chromogenic or radioactive detection methods.
  • Electrostatic, hydrophobic
    Plain/white filter
    Western blotting
    0,45 µm pore size
    Comes in an 8,5 cm × 10 m roll
The Immobilon NOW system offers 1-cut convenience for standard mini-blots (70×85 mm) and midi-blots (85×135 mm).
Compatible stains: Coomassie brilliant blue, Amido black, India ink, Ponceau-S red, Colloidal gold, CPTS, Toluidine blue, Transillumination and Sypro® ruby.