Order Entry
Austria
ContactUsLinkComponent
Mag-Bind® Endo-free Plasmid Mini Kit
Mag-Bind® Endo-free Plasmid Mini Kit
Supplier:  OMEGA BIO-TEK
New Product
Certificates
undefined
Mag-Bind® Endo-free Plasmid Mini Kit
Supplier:  OMEGA BIO-TEK
New Product

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Plasmid (endotoxin-free)
  • Sample type:
    Bacteria
  • Target molecule:
    DNA
  • Instrument Compatibility:
    MagBinder® Fit24 nucleic acid purification system

Specifications

Frequently Bought Together