Order Entry
Austria
ContactUsLinkComponent
MagBeads FastDNA™ Kit for Feces
MagBeads FastDNA™ Kit for Feces
Catalog # 116570400.
Supplier:  MP Biomedicals
CAS Number:  
undefined
MagBeads FastDNA™ Kit for Feces
Catalog # 116570400.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Description:
    MagBeads FastDNA™ kit for feces, Mini
  • Storage conditions:
    Magnetic beads should be stored at 2...8 °C, the remaining kit contents should be stored at 15...30 °C.
  • Format:
    KIT
  • Sample size:
    30 - 300 mg feces
  • Volume:
    50 preps

Specifications

About this item

The MagBeads FastDNA Kit for Feces allows thorough sample lysis and quick isolation of DNA from fecal samples. It is using magnetic beads with high DNA binding capacity for extraction of high yields of pure DNA. This is done manually using a magnetic rack or adapted to automated instruments for high throughput processing.

  • High quality DNA: Intact, high yield and free from inhibitors

  • Quick and simple: Manual extraction in 1 h
  • Wide applications: Extraction from human and animal feces, intestinal contents
  • High throughput: Compatible with automated platforms
  • Environmentally friendly: No phenol/chloroform or other toxic chemicals

The MagBeads FastDNA Kit for Feces allows quick isolation of high-quality genomic DNA from human and animal feces in less than 60 min. Samples are placed into Lysing Matrix E tube and used with FastPrep® Instruments from MP Biomedicals to effectively lyse host cells as well as bacteria, fungi, viruses, protists and other cells present in fecal samples within 40 s.

The kit is also compatible with most of the automated nucleic acid extraction instruments on the market or it can be operated manually. It consists of specially formulated reagents to eliminate humic acid, polysaccharides, phenolic compounds and enzyme inhibitors from feces and thus allow extraction of highly pure genomic DNA ready for PCR, restriction digestion, electrophoresis and other desired applications.

Target molecule: DNA

Refine target molecule: Total DNA