Order Entry
Austria
ContactUsLinkComponent
NanoDrop™ Ultra Spectrophotometers and Fluorometers
NanoDrop™ Ultra Spectrophotometers and Fluorometers
Supplier:  THERMO NANODROP
New Product
Certificates
undefined
NanoDrop™ Ultra Spectrophotometers and Fluorometers
Supplier:  THERMO NANODROP
New Product

Specifications

  • Catalog No:
  • NANDND-ULTRACSECGL
  • NANDND-ULTRA-GL
  • NANDNDU-APSEC-GL
  • NANDNDUC-APSEC-GL
  • NANDND-ULTRAC-GL
  • NANDND-ULTRA-AP-GL
  • NANDNDULTRACAPGL
  • NANDND-ULTRA-SECGL
  • Measuring modes:
  • Standard: ≤7 s, Fluorescence: ≤10 s, Acclaro Pro: ≤30 s
  • Standard: ≤7 s, Fluorescence: ≤10 s, Acclaro Pro: ≤30 s
  • Display:
  • 10 Zoll high definition colour monitor
  • 10 Zoll high definition colour monitor
  • Photometric accuracy:
  • 3% at 0,97 A, 302 nm (Extinction, expressed in abs/mm at 25 °C)
  • 3% at 0,97 A, 302 nm (Extinction, expressed in abs/mm at 25 °C)
  • Photometric range:
  • Corresponds to 10 mm: Measuring station 0 - 550 A, cuvette 0 - 1,5 A
  • Corresponds to 10 mm: Measuring station 0 - 550 A, cuvette 0 - 1,5 A
  • Wavelength range:
  • 190 - 850 nm
  • 190 - 850 nm
  • Communication interface:
  • 2×USB-A ports, 1×USB-C port, Ethernet, Bluetooth™, Wi-Fi
  • 2×USB-A ports, 1×USB-C port, Ethernet, Bluetooth™, Wi-Fi
  • Compatible with:
  • DYMO 550 Turbo printer, bluetooth keyboard, mouse and barcode reader
  • DYMO 550 Turbo printer, bluetooth keyboard, mouse and barcode reader
  • Detector:
  • Detection area: dsDNA: Pedestal: 1,0 ng/μl, Cuvette: 0,2 ng/μl, BSA (IgG): Pedestal: 0,03 (0,015) mg/ml, Cuvette: 0,006 (0,003) mg/ml; Detector type: CMOS linear image sensor with 2048 elements
  • Detection area: dsDNA: Pedestal: 1,0 ng/μl, Cuvette: 0,2 ng/μl, BSA (IgG): Pedestal: 0,03 (0,015) mg/ml, Cuvette: 0,006 (0,003) mg/ml; Detector type: CMOS linear image sensor with 2048 elements
  • Languages:
  • Chinese, English, French, German, Japanese, Spanish, Polish, Italian, Korean
  • Chinese, English, French, German, Japanese, Spanish, Polish, Italian, Korean
  • Light source:
  • Xenon flash lamp (FL version additionally with blue and red LED)
  • Xenon flash lamp (FL version additionally with blue and red LED)
  • Repeatability:
  • Typical: 0,002 A (1,0 mm path) or 1%VK, whichever is greater (SD of 10 individual measurements at 0,97 A)
  • Typical: 0,002 A (1,0 mm path) or 1%VK, whichever is greater (SD of 10 individual measurements at 0,97 A)
  • Dimensions:
  • 320L×180W×280H mm
  • 320L×180W×280H mm
  • Weight:
  • 4,1 kg
  • 4,1 kg

Specifications

Frequently Bought Together