NANOCOLOR® Photometric Tube Test Kit for Organic Acids
Catalog # MANA985050
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:Ethylenglycole/Iron(III) -Ions
- Pk:1 KIT
- Description:NANOCOLOR® organic Acids 3000, test no. 0-50
- Dimensions:160×110×250 mm (including packaging)
- Model:MANA985050
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:579.6 g (including packaging)
- For unit:UV/VIS II; VIS II; 500 D; PF-12Plus; Advance
- Tests per kit:20
- Range:30 - 3000 mg/L (CH₃COOH); 0,5 - 50,0 (mmol/L (CH₃COOH)
Specifications
About this item
Photometric test kit for the determination of organic acids in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Coloured pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery: Rugged box with 20 test tubes.