Order Entry
Austria
ContactUsLinkComponent
NANOCOLOR® Photometric Standard Test Kit for Nitrite
NANOCOLOR® Photometric Standard Test Kit for Nitrite
Catalog # MANA91867
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Standard Test Kit for Nitrite
Catalog # MANA91867
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Sulfanilic acid/1 -naphthylamine
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Nitrite, test no. 1-67
  • Dimensions:
    160×110×130 mm (including packaging)
  • Model:
    MANA91867
  • Platform:
    NANOCOLOR® standard tests
  • Storage temperature:
    15…25 °C
  • Shelf life (months):
    12
  • Weight:
    550,23 g including packaging
  • For unit:
    UV/VIS II, VIS II, Advance, 500 D, MT Easy VIS, MT Easy UV
  • Tests per kit:
    250
  • Range:
    0,002 - 0,30 mg/L (NO₂) - N; 0,005 - 1,00 mg/L (NO₂⁻)

Specifications

About this item

Photometric test kit for the determination of nitrite in water samples.

  • Ready-to-use reagents for up to 500 determinations

  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.

Sea water analysis: Yes
Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II