NANOCOLOR® Photometric Tube Tests, COD
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kits for the determination of COD in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Colored pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (AD) round cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO easy VIS and easy UV. Programming data for other spectrophotometers are available.
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No
Digestion temperature: 148 °C / 298 °F;160 °C / 320 °F
Delivery: Remark: Measuring range on NANOCOLOR VIS II
Specifications
- Model:MANA985020
- Storage temperature:2…8 °C
- Shelf life (months):12
- Weight:422,6 g (including packaging)
Specifications
Related Information
Certificates
Search by Lot NumberChemical Oxygen Demand (COD) 600 Tube Test.pdf Chemical Oxygen Demand (COD) Tube test.pdf NANOCOLOR® COD 300 Photometric Tube Tests.pdf NANOCOLOR® COD 40 Photometric Tube Tests.pdf NANOCOLOR® COD 60000 Photometric Tube Tests.pdf
Additional Product Details
More
IFU COD 60 Tube Test.pdf IFU COD-1500-HG-free.pdf IFU CSB 600 Tube Test.pdf IFU NANOCOLOR® Photometric Test Kits, COD 160 Hg‑free.pdf IFU NANOCOLOR® Photometric Test Kits, COD 300.pdf
More
Frequently Bought Together