NANOCOLOR® Photometric Standard Test Kits for Nitrate
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kits for the determination of nitrate in water samples.
- Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kits are evaluated in 10 or 50 mm rectangular cuvettes. They are pre-programmed on NANOCOLOR® photometers and are also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Sludge reagent set: No
Digestion set: No
NanOx Metal: No
NanOx N: No (except: MANA91865)
Specifications
- Model:MANA91865
- Platform:NANOCOLOR® standard tests
- Storage temperature:15...25 °C
- Shelf life (months):24
- Weight:1286 g (Including packaging)
Specifications
Related Information
Frequently Bought Together