Order Entry
Austria
ContactUsLinkComponent
NANOCOLOR® Photometric Standard Test Kit for Chloride
NANOCOLOR® Photometric Standard Test Kit for Chloride
Catalog # MANA91820
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
NANOCOLOR® Photometric Standard Test Kit for Chloride
Catalog # MANA91820
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Mercury(II) -thiocyanate/Iron(III) -nitrate
  • Pk:
    1 KIT
  • Description:
    NANOCOLOR® Chloride, test no. 1-20
  • Dimensions:
    160×110×130 mm (Including packaging)
  • Model:
    MANA91820
  • Platform:
    NANOCOLOR® standard tests
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    12
  • Weight:
    525,7 g (Including packaging)
  • For unit:
    UV/VIS II;VIS II;Advance;500 D
  • Tests per kit:
    250
  • Range:
    0,2 - 125 mg/L (Cl⁻)

Specifications

About this item

Photometric test kit for the determination of chloride in water samples.

  • Ready-to-use reagents for up to 500 determinations

  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations

The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.

Sea water analysis: No
Hazardous material: Yes

Remark: Measuring range on NANOCOLOR VIS II

Delivery:

Supplied in rugged box with reagents.