Order Entry
Austria
ContactUsLinkComponent
ReadyTube™12 MSRV Medium ISO 6579
ReadyTube™12 MSRV Medium ISO 6579
Catalog # 710-2265
Supplier:  MERCK LIFE SCIENCE
ReadyTube™12 MSRV Medium ISO 6579
Catalog # 710-2265
Supplier:  MERCK LIFE SCIENCE

About this item

  • Application: Selective enrichment for detection of Salmonella

  • Packaging: 12 ml in 17 ml tube with red cap (100 tubes per box)
  • Synonym: Modified semi-solid Rappaport-Vassiliadis Medium