Order Entry
Austria
ContactUsLinkComponent
Readyplate XLD Agar acc ISO 6579
Readyplate XLD Agar acc ISO 6579
Catalog # 710-2268
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
Readyplate XLD Agar acc ISO 6579
Catalog # 710-2268
Supplier:  MERCK LIFE SCIENCE
CAS Number:  

Specifications

  • Description:
    Readyplate XLD Agar acc ISO 6579

Specifications

About this item

  • Application: Identification of Salmonella

  • Packaging: 90 mm plate (20 plates per box)
  • Synonym: Xylose Lysine Deoxycholate Agar