NANOCOLOR® Photometric Standard Test Kit for Fluoride
Catalog # MANA918142
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:SPADNS
- Pk:1
- Description:NANOCOLOR® Fluoride, test no.1-142
- Dimensions:160×110×130 mm (including packaging)
- Seq:0
- Storage temperature:15…25 °C
- Shelf life (months):18
- Weight:565,6 g (including packaging)
- Type:Hazardous material: Yes
- For unit:UV/VIS II; VIS II; Advance; 500 D; MT Easy VIS; MT Easy UV
- Tests per kit:500
- Range:0,05 - 2,00 mg/L (F⁻)
Specifications
About this item
Photometric test kit for the determination of fluoride in water samples.
Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery: Remark: Measuring range on NANOCOLOR VIS II