NANOCOLOR® Photometric Standard Test Kit for Chromate
Catalog # 634-8124
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:Carbazide
- Pk:1 KIT
- Description:NANOCOLOR® Chromate, test no.1-25
- Dimensions:160×110×130 mm (including packaging)
- Seq:0
- Storage temperature:15…25 °C
- Shelf life (months):24
- Weight:375 g (including packaging)
- Type:Hazardous material: Yes
- For unit:UV/VIS II; VIS II; Advance; 500 D
- Tests per kit:250
- Range:0,01 - 3,0 mg/L; 0,01 - 6,0 mg/L (CrO₄²⁻)
Specifications
About this item
Photometric test kit for the determination of chromate in water samples.
Ready-to-use reagents for up to 500 determinations
- Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
- Low costs per determination
- Easy dilutions for analysis of higher concentrations
The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Delivery: Remark: Measuring range on NANOCOLOR VIS II