ContactUsLinkComponent
Steridon Sterile Coveralls with Integrated Facemask
Steridon Sterile Coveralls with Integrated Facemask
Supplier:  ISOFIELD IRELAND
New Product
Certificates
Steridon Sterile Coveralls with Integrated Facemask
Supplier:  ISOFIELD IRELAND
New Product
Specifications
  • Catalog No:
  • ISOF80231XS
  • ISOF80231XXS
  • ISOF80231L
  • ISOF80231M
  • ISOF80231S
  • ISOF80231XL
  • ISOF80231XXL
  • ISOF80231XXXL
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Cleanroom class:
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • 10/ISO 4; EU GMP (A)
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Particle shedding (Helmke Drum):
  • 160 counts/min, for particles larger than 0,3 µm
  • 160 counts/min, for particles larger than 0,3 µm
  • 160 counts/min, for particles larger than 0,3 µm
  • 160 counts/min, for particles larger than 0,3 µm
  • 160 counts/min, for particles larger than 0,3 µm
  • 160 counts/min, for particles larger than 0,3 µm
  • 160 counts/min, for particles larger than 0,3 µm
  • 160 counts/min, for particles larger than 0,3 µm