ContactUsLinkComponent
Milli-Q SQPAK™ Final Filter
Milli-Q SQPAK™ Final Filter
Catalog # MILLSQPAKFINM1
Supplier:  Merck
New Product
undefined
Milli-Q SQPAK™ Final Filter
Catalog # MILLSQPAKFINM1
Supplier:  Merck
New Product
Specifications
  • Description:
    SQPAK™ Final filter, for bacteria- and particulate-free ultrapure water for use with ultrapure dispensing module of Milli-Q® SQ 2Series systems.
Specifications
About this item
SQPAK™ Final filter ensures high-quality, bacteria- and particulate-free ultrapure (Type 1) water at the point of dispense. The purification technology includes a microfiltration filter that removes bacteria and particulates >0,22 µm.
  • Easy replacement by simply twisting
  • Replacement is recommended after 6 months
The SQPAK™ Final filter is useful for lab applications such as chemistry and microbiology.