Thermo Scientific™ TSG Undercounter Refrigerators
Supplier: Thermo Scientific
Certificates
About this item
TSG Series, powered by solid state refrigeration technology, has been designed to meet the rigorous and demanding needs of healthcare, research and industrial applications where solid performance is required.
Solid state refrigeration technology
- Whisper quiet operation leading to a more comfortable work environment supporting clinical and healthcare needs
- Increased capacities due to fewer refrigeration components means you can store more in less space
- Energy Star rated performance meaning lower operating costs
- Green refrigerants supporting global initiatives to reduce green house gas emissions
- Full alarm set, including hi/lo temperature, door ajar and power fail to support requirements for medical storage
Certified to NSF/ANSI 456 Vaccine Storage standard, the TSX series can help to meet regulations and guidelines from the CDC, World Health Organization, and other Health Ministries on vaccine storage and handling.
Ordering information:
Warranty is 5 years on the solid state refrigeration system and 2 years parts and labor.
Specifications
- Catalog No:
- HERATSG505SA
- HERATSG505GA
- Defrost:
- Automatic
- Automatic
- Shelves/drawers:
- 3 shelves
- 3 shelves
- Door type:
- 1 solid door, right hinge
- 1 glass door, right hinge
- Electrical:
- 100 - 240 V; 50 - 60 Hz
- 100 - 240 V; 50 - 60 Hz
- W×D×H ext.:
- 600×660×813 mm
- 600×660×813 mm