TriLink® Sulf Cyanine5 Aminoallyluridine-5'-Triphosphate
Supplier: TriLink BioTechnologies
New Product
Certificates
About this item
Cyanine 5-Aminoallyluridine-5'-Triphosphate is a substrate for T7 RNA polymerase, allowing the incorporation of cyanine 5 into RNA and mRNA during in vitro transcription. Cyanine 5 incorporation allows for fluorescent detection of RNA and mRNA independent of protein translation.
- Grade: RUO
Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
Frequently Bought Together