TriLink® Guanosine-5'-Triphosphate
Supplier: TriLink BioTechnologies
New Product
Certificates
About this item
This nucleotide consists of an unmodified base, a ribose, and a 5′ triphosphate group for synthesis of RNA molecules.
- Unmodified ribonucleotide for RNA synthesis
- >99% purity by AX-HPLC analysis
- Provided as 100 mM in H₂O, pH 7,5
- Grade: RUO
Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.