ContactUsLinkComponent
VWR® Premium Series Upright Ultra-Low Temperature Freezers
New Product
undefined
VWR® Premium Series Upright Ultra-Low Temperature Freezers
New Product
  • Catalog No:
  • 471-1357
  • 471-1356
  • 471-1358
  • 471-1355
  • Model:
  • Premium ULT freezer 600
  • Premium ULT freezer 500
  • Premium ULT freezer 700
  • Premium ULT freezer 400
  • Cat. No.:
  • 471-1357
  • 471-1356
  • 471-1358
  • 471-1355
  • Electrical:
  • 230 V, 16A
  • 230 V, 16A
  • 230 V, 16A
  • 230 V, 16A
  • Shelves/drawers:
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • 4 shelves (3 adjustable)
  • Door type:
  • 1 outer, 2 inner
  • 1 outer, 2 inner
  • 1 outer, 2 inner
  • 1 outer, 2 inner
  • Door lock:
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • Key lock with padlock hasps
  • W×D×H int.:
  • 874×719×1301 mm
  • 732×719×1301 mm
  • 1019×719×1301 mm
  • 588×719×1301 mm
  • W×D×H ext.:
  • 1006×950×1981 mm
  • 864×950×1981 mm
  • 1148×950×1981 mm
  • 719×950×1981 mm
  • Weight:
  • 327 kg
  • 297 kg
  • 347 kg
  • 270 kg
Replacement Parts
Frequently Bought Together