Order Entry
Austria
Orders LinkContactUsLinkComponent
cAMP ELISA Kit

cAMP ELISA Kit

New Product
Supplier:   Antibodies.com
cAMP ELISA Kit
Supplier:  Antibodies.com
New Product
Certificates

2 Options Available Below

Product Details & Documents

cAMP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of cAMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids.

cAMP ELISA Kit employs the competitive enzyme immunoassay technique for the quantitative measurement of cAMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. The 96-well microtiter plate has been pre-coated with cAMP antigen. During the incubation, cAMP present in the samples or standards competes with the fixed amount of immobilized cAMP for binding sites on the Biotinylated Anti-cAMP Antibody. The more cAMP present in a sample or standard, the less Biotinylated Anti-cAMP Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-cAMP Antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of cAMP present in each sample or standard. The concentration of cAMP can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ANTIA1641-48
  • ANTIA1641-96
  • Target protein:
  • cAMP
  • cAMP
  • Assay Type:
  • Competitive
  • Competitive
  • Format:
  • Pre-coated
  • Pre-coated
  • Assay duration:
  • Multiple steps
  • Multiple steps
  • Sensitivity:
  • 0,118 ng/ml
  • 0,118 ng/ml
  • Detection range:
  • 0,312 - 20 ng/ml
  • 0,312 - 20 ng/ml
  • Detection method:
  • Colorimetric
  • Colorimetric
  • Detection limit:
  • Intra-assay: CV <10%, Inter-assay: CV <12%
  • Intra-assay: CV <10%, Inter-assay: CV <12%
  • Sample type:
  • Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
  • Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
  • Time to Results:
  • 2 h 30 min
  • 2 h 30 min
  • Regulatory status:
  • RUO
  • RUO
  • Storage temperature:
  • 4 °C
  • 4 °C
  • Shelf life:
  • Store for 6 months at 4 °C
  • Store for 6 months at 4 °C

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • ANTIA1641-48
    cAMP ELISA kit
    cAMP ELISA kit
    Log in for availability
    Log in for availability
    1 * 48 Tests
    1 * 48 Tests - 490,00 €
    490,00 €
    1 * 48 Tests
       490,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ANTIA1641-48
    Target protein
    cAMP
    Assay Type
    Competitive
    Format
    Pre-coated
    Assay duration
    Multiple steps
    Sensitivity
    0,118 ng/ml
    Detection range
    0,312 - 20 ng/ml
    Detection method
    Colorimetric
    Detection limit
    Intra-assay: CV <10%, Inter-assay: CV <12%
    Sample type
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
    Time to Results
    2 h 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
    Shelf life
    Store for 6 months at 4 °C
    New Product
  • ANTIA1641-96
    cAMP ELISA kit
    cAMP ELISA kit
    Log in for availability
    Log in for availability
    1 * 96 Tests
    1 * 96 Tests - 645,00 €
    645,00 €
    1 * 96 Tests
       645,00 €

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ANTIA1641-96
    Target protein
    cAMP
    Assay Type
    Competitive
    Format
    Pre-coated
    Assay duration
    Multiple steps
    Sensitivity
    0,118 ng/ml
    Detection range
    0,312 - 20 ng/ml
    Detection method
    Colorimetric
    Detection limit
    Intra-assay: CV <10%, Inter-assay: CV <12%
    Sample type
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids
    Time to Results
    2 h 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
    Shelf life
    Store for 6 months at 4 °C
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.