Mouse Complement C5 ELISA Kit
Mouse Complement C5 ELISA kit, 96 tests
Catalog # ABCAAB264609-1X96
Supplier: Abcam
Jump to:
Product Details & Documents
Mouse Complement C5 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Complement C5 with a sensitivity of 1,97 ng/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.
Specifications
Specifications
- Description:Mouse Complement C5 ELISA kit
- Tests per kit:96 tests
- Cat. No.:ABCAAB264609-1X96
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:1,97 ng/ml
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Sample type:Cell culture media, EDTA Plasma, Heparin Plasma, Serum
- Target protein:C5
- Detection range:4,69 - 300 ng/ml
- Storage temperature:+4 °C
- Regulatory status:RUO
- Assay Type:Sandwich



