Human Cathepsin E ELISA Kit
Human Cathepsin E ELISA kit, 96 tests
ABCAAB313522-96TST
: Abcam
Human Cathepsin E ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Cathepsin E in Human in cell culture supernatant, serum samples.
- Description:Human Cathepsin E ELISA kit
- Tests per kit:96 tests
- Cat. No.:ABCAAB313522-96TST
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:14,5 pg/ml
- Format:Pre-coated
- Primary antibody reactivity:Human
- Sample type:Cell culture supernatant, Serum
- Target protein:Cathepsin E
- Detection range:14,5 - 4000 pg/ml
- Storage temperature:4 ℃
- Regulatory status:RUO
- Assay Type:Sandwich
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



