Order Entry
Austria
ContactUsLinkComponent
HRP Stability Conjugate Reagent
HRP Stability Conjugate Reagent
Supplier:  Abcam
Certificates
undefined
HRP Stability Conjugate Reagent
Supplier:  Abcam

Specifications

  • Form:
    Liquid
  • Storage temperature:
    Store at room temperature
  • Application:
    ELISA buffer

Specifications

Frequently Bought Together