Order Entry
Austria
Orders LinkContactUsLinkComponent
Human IL-10 ELISPOT Set

Human IL-10 ELISPOT Set

Supplier:   Abcam
Human IL-10 ELISPOT Set
Supplier:  Abcam
Certificates
Human IL-10 ELISPOT Set

4 Options Available Below

Product Details & Documents

Human IL-10 ELISPOT Set is a ELISPOT Set for the sensitive detection of Human IL-10 production in a single cell suspension.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB46549-480
  • ABCAAB46549-10X96
  • ABCAAB46549-15X96
  • ABCAAB46549-20X96
  • Antigen symbol:
  • IL-10
  • IL-10
  • IL-10
  • IL-10
  • Form:
  • Uncoated
  • Uncoated
  • Uncoated
  • Uncoated
  • Processing time:
  • Multiple steps
  • Multiple steps
  • Multiple steps
  • Multiple steps
  • Regulatory status:
  • RUO
  • RUO
  • RUO
  • RUO
  • Sample type:
  • Suspension cells
  • Suspension cells
  • Suspension cells
  • Suspension cells
  • Species reactivity:
  • Human
  • Human
  • Human
  • Human
  • Storage temperature:
  • 4 °C
  • 4 °C
  • 4 °C
  • 4 °C

Specifications

Product Family Options

Product Information

  • Availability
    Price
  • ABCAAB46549-480
    Human IL-10 ELISPOT Set
    Human IL-10 ELISPOT Set
    Log in for availability
    Log in for availability
    1 * 480 Tests
    1 * 480 Tests - 1 110,00 €
    1 110,00 €
    1 * 480 Tests
       1 110,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB46549-480
    Antigen symbol
    IL-10
    Form
    Uncoated
    Processing time
    Multiple steps
    Regulatory status
    RUO
    Sample type
    Suspension cells
    Species reactivity
    Human
    Storage temperature
    4 °C
  • ABCAAB46549-10X96
    Human IL-10 ELISPOT Set
    Human IL-10 ELISPOT Set
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 1 700,00 €
    1 700,00 €
    1 * 1 KIT
       1 700,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB46549-10X96
    Antigen symbol
    IL-10
    Form
    Uncoated
    Processing time
    Multiple steps
    Regulatory status
    RUO
    Sample type
    Suspension cells
    Species reactivity
    Human
    Storage temperature
    4 °C
  • ABCAAB46549-15X96
    Human IL-10 ELISPOT Set
    Human IL-10 ELISPOT Set
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 2 350,00 €
    2 350,00 €
    1 * 1 KIT
       2 350,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB46549-15X96
    Antigen symbol
    IL-10
    Form
    Uncoated
    Processing time
    Multiple steps
    Regulatory status
    RUO
    Sample type
    Suspension cells
    Species reactivity
    Human
    Storage temperature
    4 °C
  • ABCAAB46549-20X96
    Human IL-10 ELISPOT Set
    Human IL-10 ELISPOT Set
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 2 790,00 €
    2 790,00 €
    1 * 1 KIT
       2 790,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB46549-20X96
    Antigen symbol
    IL-10
    Form
    Uncoated
    Processing time
    Multiple steps
    Regulatory status
    RUO
    Sample type
    Suspension cells
    Species reactivity
    Human
    Storage temperature
    4 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.