Order Entry
Austria
Orders LinkContactUsLinkComponent
Human PAP ELISA Kit (ACPP)

Human PAP ELISA Kit (ACPP)

Human PAP ELISA Kit (ACPP), 96 tests
Catalog # ABCAAB267802-1X96
Supplier:   Abcam
Human PAP ELISA Kit (ACPP)
Catalog # ABCAAB267802-1X96
Supplier:  Abcam
CAS Number:  
Human PAP ELISA Kit (ACPP)
Human PAP ELISA Kit (ACPP)

Human PAP ELISA Kit (ACPP), 96 tests

CAS Number:  
Price
795,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Human PAP ELISA kit (ACPP) is a single-wash 90-min Simplestep used to quantify Human PAP (ACPP) with a sensitivity of 9.73 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human PAP ELISA Kit (ACPP)
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Urine, Cell culture media, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
  • Detection range:
    37,5 - 2400 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 hr 30 min
  • Cat. No.:
    ABCAAB267802-1X96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    9,73 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    PAP
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.