Anti-HLA-DPB1 Rabbit Monoclonal Antibody [clone: EPR11226] (Alexa Fluor® 488)
ABCAAB201527-100
: Abcam
- Seq:1286
- Size:100 µl
- Pack type:Vial
Alexa Fluor® 488 Rabbit monoclonal [EPR11226] to HLA-DPB1.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: EPR11226
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human