Anti-Nucleoprotein Crimean-Congo hemorrhagic fever virus Rabbit Polyclonal Antibody
Catalog # ABCAAB190657-50
Supplier: Abcam
Specifications
- Size:50 µg
- Pack type:Vial
Specifications
About this item
Rabbit polyclonal to Nucleoprotein Crimean-Congo hemorrhagic fever virus.
Type: Primary
Antigen: Nucleoprotein Crimean-Congo hemorrhagic fever virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: CCHFV