Anti-BNIP3 Mouse Monoclonal Antibody [clone: ANa40] (HRP (Horseradish Peroxidase))
ABCAAB196707-100
: Abcam
- Seq:1286
- Size:100 µl
- Pack type:Vial
HRP Mouse monoclonal [ANa40] to BNIP3.
: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: BNIP3
Clonality: Monoclonal
Clone: ANa40
Conjugation: HRP (Horseradish Peroxidase)
Epitope: The epitope recognized by the antibody resides within amino acids 112-124 of human BNIP3 molecule
Host: Mouse
Isotype: IgG2b
Reactivity: Human