Order Entry
Austria
ContactUsLinkComponent
Rat Total Superoxide Dismutase ELISA Kit
Rat Total Superoxide Dismutase ELISA Kit
Catalog # ANTIA319446-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
Rat Total Superoxide Dismutase ELISA Kit
Catalog # ANTIA319446-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Total Superoxide Dismutase
  • Description:
    Rat Total Superoxide Dismutase ELISA kit
  • Environmentally Preferable:
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Rat Total Superoxide Dismutase ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Total Superoxide Dismutase. No significant cross-reactivity or interference between Total Superoxide Dismutase and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 hours
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,781 - 50 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    100 μl
  • Sensitivity:
    0,469 ng/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Rat Total Superoxide Dismutase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Total Superoxide Dismutase in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Rat Total Superoxide Dismutase ELISA Kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Total Superoxide Dismutase in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Total Superoxide Dismutase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Total Superoxide Dismutase present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Total Superoxide Dismutase Antibody, which binds the captured Total Superoxide Dismutase present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Total Superoxide Dismutase captured in each well. The concentration of Total Superoxide Dismutase can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Delivery: This is a Ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch and protocol manual.
Caution: This product is for Research use only. It is not for use in diagnostic or therapeutic procedures.