Orders LinkContactUsLinkComponent
Capillary column splitter kits
Capillary column splitter kits
Supplier:   Agilent
Capillary column splitter kits
Supplier:  Agilent
Certificates
Capillary column splitter kits
3 Options Available Below
Product Details & Documents
These splitters are used for splitting an injected sample from a single injector into two columns and on to two detectors.
The dimensions of both the columns, length and internal diameter (ID), should be the same so that the vaporized sample will get spilt into almost equal amount on to two columns.
Delivery information: Kit which comes with transfer line, splitter union and necessary ferrules.
Product Family Options
Product Information
  • Ø int.
    Availability
    Price
  • HEWL5181-3390
    Fixed inlet splitter kit
    Fixed inlet splitter kit
    0,22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 645,00 €
    645,00 €
    1 * 1 ST
       645,00 €
  • HEWL5181-3391
    Fixed inlet splitter kit
    Fixed inlet splitter kit
    0,32 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 646,00 €
    646,00 €
    1 * 1 ST
       646,00 €
  • HEWL5181-3392
    Fixed inlet splitter kit
    Fixed inlet splitter kit
    0,53 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 646,00 €
    646,00 €
    1 * 1 ST
       646,00 €