Orders LinkContactUsLinkComponent
Micro filter funnels
Micro filter funnels
Supplier:   SciLabware
Micro filter funnels
Supplier:  SciLabware
Certificates
Micro filter funnels
Micro filter funnels
2 Options Available Below
Product Details & Documents
Micro funnels, with sintered disc, made of PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for filtering small amounts of material.
Product Family Options
Product Information
  • Plate Ø
    Porosity
    Capacity
    Stem O-Ø
    Availability
    Price
  • 15 mm
    1
    8 ml
    8 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 504,00 €
    504,00 €
    1 * 20 ST
       504,00 €
  • 15 mm
    2
    8 ml
    8 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 252,00 €
    252,00 €
    1 * 10 ST
       252,00 €