Orders LinkContactUsLinkComponent
Filtration funnels
Filtration funnels
Supplier:   SciLabware
Filtration funnels
Supplier:  SciLabware
Certificates
Filtration funnels
Filtration funnels
3 Options Available Below
Product Details & Documents
Pyrex® borosilicate glass.
  • Sintered glass disc
  • Filter paper is not required for filtration of solutions
  • Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
Product Family Options
Product Information
  • Capacity
    Length
    Stem length
    Porosity
    Stem O-Ø
    Int.Ø top
    Availability
    Price
  • 20 ml
    130 mm
    75 mm
    3
    8 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 198,00 €
    198,00 €
    1 * 5 ST
       198,00 €
  • 20 ml
    130 mm
    75 mm
    4
    8 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 219,00 €
    219,00 €
    1 * 5 ST
       219,00 €
  • 30 ml
    144 mm
    82 mm
    3
    8 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,20 €
    57,20 €
    1 * 1 ST
       57,20 €