Orders LinkContactUsLinkComponent
Sample cuvettes
 :   Hach
Sample cuvettes
 :  Hach
Sample cuvettes
Sample cuvettes
These high range sample cuvettes are designed for use with all spectrophotometers.
Suitable for chlorine applications.
Product Family Options
  • Description
    Ext.Ø
    Height
    Volume
    Path length
    Window material
  • Sample cell
    25 mm
    60 mm
    10 ml
    25,4 mm
    Glass
    1 * 6 ST
    1 * 6 ST - 119,00 €
    119,00 €
    1 * 6 ST
       119,00 €
  • Sample cell, with cap
    25 mm
    60 mm
    10 ml
    10 mm
    PS
    1 * 2 ST
    1 * 2 ST - 47,90 €
    47,90 €
    1 * 2 ST
       47,90 €
  • Sample cell
    25 mm
    60 mm
    5/10 ml
    10 mm
    Plastic
    1 * 2 ST
    1 * 2 ST - 42,00 €
    42,00 €
    1 * 2 ST
       42,00 €