Orders LinkContactUsLinkComponent
Calibration weights, cylindrical, OIML, class E2
Calibration weights, cylindrical, OIML, class E2
Supplier:   Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Supplier:  Sartorius Balances
Certificates
Calibration weights, cylindrical, OIML, class E2
Calibration weights, cylindrical, OIML, class E2
21 Options Available Below
Product Details & Documents
Class E2 cyclindrical calibration weights.
Product Family Options
E2 weights in plastic cases without certificate
Product Information
  • Weight
    Tolerance
    Type
    Availability
    Price
  • 1 g
    0,030 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,70 €
    84,70 €
    1 * 1 ST
       84,70 €
  • 2 g
    0,040 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,20 €
    93,20 €
    1 * 1 ST
       93,20 €
  • 5 g
    0,050 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,60 €
    98,60 €
    1 * 1 ST
       98,60 €
  • 10 g
    0,060 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
  • 20 g
    0,080 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 50 g
    0,10 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 135,00 €
    135,00 €
    1 * 1 ST
       135,00 €
  • 100 g
    0,16 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 160,00 €
    160,00 €
    1 * 1 ST
       160,00 €
  • 200 g
    0,30 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
  • 500 g
    0,8 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 314,00 €
    314,00 €
    1 * 1 ST
       314,00 €
  • 1 kg
    1,6 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 471,00 €
    471,00 €
    1 * 1 ST
       471,00 €
E2 weights in wooden boxes without certificate
Product Information
  • Weight
    Tolerance
    Type
    Availability
    Price
  • 2 kg
    3,0 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 724,00 €
    724,00 €
    1 * 1 ST
       724,00 €
  • 5 kg
    8,0 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 060,00 €
    1 060,00 €
    1 * 1 ST
       1 060,00 €
E2 weights in plastic cases with DAkkS certificate
Product Information
  • Weight
    Tolerance
    Type
    Availability
    Price
  • 1 g
    0,030 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 172,00 €
    172,00 €
    1 * 1 ST
       172,00 €
  • 5 g
    0,050 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 186,00 €
    186,00 €
    1 * 1 ST
       186,00 €
  • 100 g
    0,16 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 248,00 €
    248,00 €
    1 * 1 ST
       248,00 €
  • 200 g
    0,30 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 286,00 €
    286,00 €
    1 * 1 ST
       286,00 €
  • 500 g
    0,8 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 403,00 €
    403,00 €
    1 * 1 ST
       403,00 €
  • 1 kg
    1,6 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 558,00 €
    558,00 €
    1 * 1 ST
       558,00 €
E2 weights in wooden boxes with DAkkS certificate
Product Information
  • Weight
    Tolerance
    Type
    Availability
    Price
  • 50 g
    0,10 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 221,00 €
    221,00 €
    1 * 1 ST
       221,00 €
  • 2 kg
    3,0 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 811,00 €
    811,00 €
    1 * 1 ST
       811,00 €
  • 5 kg
    8,0 ± mg
    Cylindrical test weight
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 150,00 €
    1 150,00 €
    1 * 1 ST
       1 150,00 €