Microtubes, Prepared
Marketsource
Supplier: SARSTEDT
Jump to:
About this item
PP, translucent with PP screw cap.
- Type of preparation: Trisodium citrate solution
- With paper label
- Graduated
- Recommended centrifugation: 1800×g
With screw cap or push-on cap.
15 Options Available Below
Product Details & Documents
PP, translucent with PP screw cap.
- Type of preparation: Trisodium citrate solution
- With paper label
- Graduated
- Recommended centrifugation: 1800×g
With screw cap or push-on cap.
Product Family Options
With screw cap
Product Information
- CapacityPackedAvailabilityPrice
- SARS41.1378.005Microtubes, prepared, serum-gel clotting activator, with screw cap closureMicrotubes, prepared, serum-gel clotting activator, with screw cap closure1,1 ml1000/caseLog in for availability1 * 1.000 ST334,00 €Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- SARS41.1350.005Microtubes, prepared, coagulation citrate, with screw cap closureMicrotubes, prepared, coagulation citrate, with screw cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST188,00 €Specifications:MarketsourceMarketsource
- SARS41.1392.005Microtubes, prepared, serum clotting activator, with screw cap closureMicrotubes, prepared, serum clotting activator, with screw cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST148,00 €Specifications:MarketsourceMarketsource
- SARS41.1393.005Microtubes, prepared, plasma-lithium heparin, with screw cap closureMicrotubes, prepared, plasma-lithium heparin, with screw cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST193,00 €Specifications:MarketsourceMarketsource
- SARS41.1394.005Microtubes, prepared, glucose fluoride, with screw cap closureMicrotubes, prepared, glucose fluoride, with screw cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST148,00 €Specifications:MarketsourceMarketsource
- SARS41.1395.100Microtubes, prepared, haematology potassium EDTA, with screw cap closureMicrotubes, prepared, haematology potassium EDTA, with screw cap closure1,3 ml100/caseLog in for availability1 * 100 STSpecifications:MarketsourceMarketsource
With push cap
Product Information
- CapacityPackedAvailabilityPrice
- SARS41.1505.005Microtubes, prepared, glucose fluoride, with attached push cap closureMicrotubes, prepared, glucose fluoride, with attached push cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST148,00 €Specifications:MarketsourceMarketsource
- SARS41.1503.005Microtubes, prepared, plasma-lithium heparin, with attached push cap closureMicrotubes, prepared, plasma-lithium heparin, with attached push cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST193,00 €Specifications:MarketsourceMarketsource
- SARS41.1504.005Microtubes, prepared, haematology potassium EDTA, with attached push cap closureMicrotubes, prepared, haematology potassium EDTA, with attached push cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST193,00 €Specifications:MarketsourceMarketsource
- SARS41.1500.005Microtubes, prepared, serum-gel clotting activator, with attached push cap closureMicrotubes, prepared, serum-gel clotting activator, with attached push cap closure1,1 ml1000/caseLog in for availability1 * 1.000 ST334,00 €Specifications:MarketsourceMarketsource
- SARS41.1501.005Microtubes, prepared, serum clotting activator, with attached push cap closureMicrotubes, prepared, serum clotting activator, with attached push cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST193,00 €Specifications:MarketsourceMarketsource
- SARS41.1506Microtubes, prepared, coagulation citrate, with attached push cap closureMicrotubes, prepared, coagulation citrate, with attached push cap closure1,0 ml1000/caseLog in for availability1 * 1.000 ST188,00 €Specifications:MarketsourceMarketsource
- SARS41.1506.002Microtubes, prepared, coagulation citrate, with attached push cap closureMicrotubes, prepared, coagulation citrate, with attached push cap closure0,5 ml1000/caseLog in for availability1 * 1.000 ST202,00 €Specifications:MarketsourceMarketsource
- SARS41.1506.005Microtubes, prepared, coagulation citrate, with attached push cap closureMicrotubes, prepared, coagulation citrate, with attached push cap closure1,3 ml1000/caseLog in for availability1 * 1.000 ST188,00 €Specifications:MarketsourceMarketsource