Empty solid phase extraction cartridges, Bond Elut™
Anbieter: VARIAN
Jump to:
About this item
Bond Elut empty SPE cartridges are made with high purity polypropylene for cleaner extracts.
- Uniform batch-to-batch size for consistent performance
- Cartridges are available from 1 to 60 ml
Ordering information: Frits has to be ordered separately
6 Options Available Below
Product Details & Documents
Bond Elut empty SPE cartridges are made with high purity polypropylene for cleaner extracts.
- Uniform batch-to-batch size for consistent performance
- Cartridges are available from 1 to 60 ml
Ordering information: Frits has to be ordered separately
Product Family Options
Product Information
- Column FormatCapacityVerfügbarkeitPreis
Accessories
Product Information
- Artikelnr.DescriptionVerfügbarkeitPreis
Product Information
- Artikelnr.DescriptionVerfügbarkeitPreis
Product Information
- Artikelnr.DescriptionVerfügbarkeitPreis
- VARI12131035Adapter caps for3 ml Gilson ASPEC SPE systemsLog in for availability1 * 1.000 ST - 1 043,46 €
Product Information
- Artikelnr.DescriptionVerfügbarkeitPreis
- VARI12131034Adapter caps for1ml Gilson ASPEC SPE systemsLog in for availability1 * 1.000 ST - 824,54 €