Order Entry
Austria
ContactUsLinkComponent
Roux culture flasks, PYREX®
Roux culture flasks, PYREX®
Catalog # 392-0361
Supplier:  SciLabware
undefined
Roux culture flasks, PYREX®
Catalog # 392-0361
Supplier:  SciLabware

Specifications

  • Capacity:
    1200 ml
  • Height:
    123 mm
  • Seq:
    03
  • Pk:
    10

Specifications

About this item

Pyrex® borosilicate glass.

  • Ideal for cell culture
  • Chemically inert glass for a contamination-free environment
  • Off-set neck