Cartridge burner, high temperature
USBE1424
- Max. temp.:1600 °C
- Description:High temperature burner, without cartridge
- Environmentally Preferable:
- Pk:1
High temperature burner with piezo electric ignition and needle valve. Maximum temperature of 1600 °C.