ContactUsLinkComponent
Micro burner with needle valve, cartridge type
Micro burner with needle valve, cartridge type
Catalog # USBE1423
undefined
Micro burner with needle valve, cartridge type
Catalog # USBE1423
Specifications
  • Max. temp.:
    1640 °C
  • Description:
    Micro burner, without cartridge
  • Pk:
    1
Specifications
About this item
Micro burner with piezo electric ignition and air regulation. Maximum temperature of 1640 °C.
Cartridges
Product Information