ContactUsLinkComponent
Spreaders, T-shaped
Spreaders, T-shaped
Catalog # SARS86.1569.005
Supplier:  SARSTEDT
Spreaders, T-shaped
Catalog # SARS86.1569.005
Supplier:  SARSTEDT
Specifications
  • Packed:
    4/peel pack
  • Colour:
    Blue
  • Description:
    Innoculation spreader, PS
  • Pk:
    500
Specifications
About this item
Sterile inoculation spreaders made from blue PS. Ideal for spreading a bacterial suspension.