Desiccators, PYREX®
Supplier: SciLabware
Jump to:
About this item
Ideal for vacuum and non vacuum applications.
- Ground glass flanges
- Borosilicate glass base and lid
- Plates positioned on an internal ledge within the base
5 Options Available Below
Product Details & Documents
Ideal for vacuum and non vacuum applications.
- Ground glass flanges
- Borosilicate glass base and lid
- Plates positioned on an internal ledge within the base
Product Family Options
Non-vacuum type
Product Information
- TypeNominal sizeUsable heightØ ext.×HØ int.AvailabilityPrice
- BIBB1590/02DDesiccatorDesiccatorNon-vacuum, for use at atmospheric pressure, with knob150 DN85 mm215×154 mm170 mmLog in for availability1 * 1 ST200,00 €
- BIBB1591/02DDesiccatorDesiccatorNon-vacuum, for use at atmospheric pressure, with knob200 DN120 mm270×202 mm220 mmLog in for availability1 * 1 ST222,00 €
Vacuum type
Product Information
- TypeNominal sizeUsable heightØ ext.×HØ int.AvailabilityPrice
- BIBB1593/02DDesiccatorDesiccatorFor use under vacuum, with 24/29 socket and stopcock150 DN85 mm215×154 mm170 mmLog in for availability1 * 1 ST253,00 €
- BIBB1594/02DDesiccatorDesiccatorFor use under vacuum, with 24/29 socket and stopcock200 DN120 mm270×202 mm220 mmLog in for availability1 * 1 ST300,00 €
Stopcocks
Product Information
- TypeNominal sizeUsable heightØ ext.×HØ int.AvailabilityPrice
- BIBB1612/03Rotaflo® stopcock with 24/29 joint for vacuum tight seal
Desiccator bases
Product Information
- Catalog #Usable heightNominal sizeDescriptionTypeØ ext.×HAvailabilityPrice
- 85 mm150 DNPYREX® borosilicate glass baseWith flat flange Ø int. 170 mm215×154 mmLog in for availability1 * 1 ST173,00 €
Spare lids
Product Information
- Catalog #Nominal sizeDescriptionTypeAvailabilityPrice
- 200 DNPYREX® borosilicate glass lidFor desiccators non vacuum typeLog in for availability1 * 1 ST106,00 €
Desiccator plates
Product Information
- Catalog #ØDescriptionTypeAvailabilityPrice
- 150 mmMetal perforated platePyrex® vacuum and non-vacuum desiccatorsLog in for availability1 * 1 ST24,40 €